The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_003499743.2 | XP_003499791.1 | 2 - 3 | 1042 | 68 |
GenBank mRNA | JP052484.1 | - | 2 - 3 | 430 | 68 |