Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Il27
Entrez Gene ID
100753480
Gene Name
interleukin 27
Gene Aliases
I79_024209
Chromosome Location
Chr.Un|NW_003617044: 41034 - 45955 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003514857.2 XP_003514905.1 3 - 4 293 87

View Full Product Details