Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Csdc2
Entrez Gene ID
100759782
Gene Name
cold shock domain containing C2, RNA binding
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003616508: 106238 - 115383 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003514312.2 XP_003514360.2 3 - 4 547 75

View Full Product Details