Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ovol1
Entrez Gene ID
100760732
Gene Name
ovo-like zinc finger 1
Gene Aliases
I79_021808
Chromosome Location
Chr.Un|NW_003615656: 69152 - 80078 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007653323.1 XP_007651513.1 7 - 8 1334 92

View Full Product Details