Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ccdc71
Entrez Gene ID
100762268
Gene Name
coiled-coil domain containing 71
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615379: 2002 - 6437 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512159.2 XP_003512207.1 2 - 2 597 72
XM_007652643.1 XP_007650833.1 2 - 2 639 72

View Full Product Details