Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Heatr2
Entrez Gene ID
100771212
Gene Name
HEAT repeat containing 2
Gene Aliases
I79_014666
Chromosome Location
Chr.Un|NW_003614324: 354603 - 395380 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003507074.2 XP_003507122.1 12 - 13 2004 57
GenBank mRNA JI889071.1 - 1 - 2 163 57

View Full Product Details