Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Iba57
Entrez Gene ID
100763588
Gene Name
IBA57, iron-sulfur cluster assembly homolog (S. cerevisiae)
Gene Aliases
I79_010130
Chromosome Location
Chr.Un|NW_003613960: 401667 - 409600 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007643941.1 XP_007642131.1 1 - 2 347 117

View Full Product Details