Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fbxo39
Entrez Gene ID
100754321
Gene Name
F-box protein 39
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614234: 621969 - 624299 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506364.1 XP_003506412.1 1 - 2 1023 74

View Full Product Details