Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mmrn2
Entrez Gene ID
100774816
Gene Name
multimerin 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613589: 3960041 - 3978608 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495326.1 XP_003495374.1 4 - 5 486 68

View Full Product Details