Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Btla
Entrez Gene ID
100771853
Gene Name
B and T lymphocyte associated
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613667: 909567 - 937928 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007619078.1 XP_007617268.1 4 - 5 581 82

View Full Product Details