Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prok2
Entrez Gene ID
100759658
Gene Name
prokineticin 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613585: 2391216 - 2510891 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007620746.1 XP_007618936.1 4 - 5 286 66

View Full Product Details