Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sgcz
Entrez Gene ID
100773002
Gene Name
sarcoglycan, zeta
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613671: 2297987 - 2584228 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003497954.2 XP_003498002.1 6 - 7 712 72

View Full Product Details