Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNRPG
Entrez Gene ID
102190526
Gene Name
small nuclear ribonucleoprotein polypeptide G
Gene Aliases
-
Chromosome Location
Chr.11: 68326090 - 68334650 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005686845.3 XP_005686902.1 3 - 4 278 80

View Full Product Details