Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBRG4
Entrez Gene ID
102181265
Gene Name
transforming growth factor beta regulator 4
Gene Aliases
-
Chromosome Location
Chr.4: 43648243 - 43658631 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005679316.3 XP_005679373.1 10 - 11 1946 64
XM_013963385.2 XP_013818839.1 10 - 11 1863 64
GenBank mRNA JO542380.2 - 1 - 2 394 64

View Full Product Details