Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NPAS4
Entrez Gene ID
102176203
Gene Name
neuronal PAS domain protein 4
Gene Aliases
-
Chromosome Location
Chr.29: 44865243 - 44870838 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005699921.2 XP_005699978.1 1 - 2 287 108

View Full Product Details