Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ICAM4
Entrez Gene ID
102187395
Gene Name
intercellular adhesion molecule 4 (Landsteiner-Wiener blood group)
Gene Aliases
-
Chromosome Location
Chr.7: 94708511 - 94709659 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018051025.1 XP_017906514.1 1 - 2 377 58

View Full Product Details