Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNBD1
Entrez Gene ID
102173401
Gene Name
cyclic nucleotide binding domain containing 1
Gene Aliases
-
Chromosome Location
Chr.14: 6412362 - 6818037 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018058081.1 XP_017913570.1 1 - 2 82 105

View Full Product Details