Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRPL10
Entrez Gene ID
102172754
Gene Name
mitochondrial ribosomal protein L10
Gene Aliases
-
Chromosome Location
Chr.19: 38259810 - 38265855 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005693717.3 XP_005693774.2 1 - 2 112 83

View Full Product Details