Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLCXD3
Entrez Gene ID
102179239
Gene Name
phosphatidylinositol specific phospholipase C X domain containing 3
Gene Aliases
-
Chromosome Location
Chr.20: 33010352 - 33209619 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005694750.3 XP_005694807.1 2 - 3 1023 66

View Full Product Details