Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRIM65
Entrez Gene ID
102171278
Gene Name
tripartite motif containing 65
Gene Aliases
-
Chromosome Location
Chr.19: 54953275 - 54958261 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018063783.1 XP_017919272.1 5 - 6 998 72

View Full Product Details