Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ERMN
Entrez Gene ID
102175811
Gene Name
ermin
Gene Aliases
-
Chromosome Location
Chr.2: 97130254 - 97137920 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005676116.3 XP_005676173.2 2 - 3 667 95
GenBank mRNA JO634162.2 - 1 - 2 3122 95
JT725554.1 - 3 - 4 628 95

View Full Product Details