Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTC32
Entrez Gene ID
102176891
Gene Name
tetratricopeptide repeat domain 32
Gene Aliases
-
Chromosome Location
Chr.11: 78628315 - 78636944 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005686979.3 XP_005687036.2 1 - 2 764 109

View Full Product Details