Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PCNP
Entrez Gene ID
102169907
Gene Name
PEST proteolytic signal containing nuclear protein
Gene Aliases
-
Chromosome Location
Chr.1: 45740750 - 45755856 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013975550.2 XP_013831004.2 1 - 2 463 107
GenBank mRNA JO324225.2 - 4 - 5 596 107
JO363573.2 - 1 - 2 118 107
JO394550.2 - 4 - 5 768 107
JO451355.2 - 4 - 5 588 107
JO498946.2 - - 329 107
JO556981.2 - 1 - 2 148 107
JO556982.2 - 1 - 2 148 107
JO610616.2 - 4 - 5 587 107
KA310411.1 - 4 - 5 1291 107

View Full Product Details