Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IL27
Entrez Gene ID
102183199
Gene Name
interleukin 27
Gene Aliases
-
Chromosome Location
Chr.25: 25969955 - 25975455 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005697655.3 XP_005697712.2 4 - 5 645 66

View Full Product Details