Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HIGD1B
Entrez Gene ID
102186048
Gene Name
HIG1 hypoxia inducible domain family member 1B
Gene Aliases
-
Chromosome Location
Chr.19: 44200484 - 44203011 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005693939.3 XP_005693996.1 2 - 3 333 91
XM_005693940.3 XP_005693997.1 2 - 3 273 91
GenBank mRNA JO643619.2 - 2 - 3 278 91
KA332385.1 - 2 - 3 267 91

View Full Product Details