Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HIGD1B
Entrez Gene ID
102186048
Gene Name
HIG1 hypoxia inducible domain family member 1B
Gene Aliases
-
Chromosome Location
Chr.19: 44200484 - 44203011 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005693939.3 XP_005693996.1 3 - 4 464 117
XM_005693940.3 XP_005693997.1 3 - 4 404 117
GenBank mRNA JO643619.2 - 1 - 2 147 117
KA332385.1 - 1 - 2 136 117

View Full Product Details