Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PEX11A
Entrez Gene ID
102190821
Gene Name
peroxisomal biogenesis factor 11 alpha
Gene Aliases
-
Chromosome Location
Chr.21: 20395680 - 20402525 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005694973.3 XP_005695030.1 2 - 3 303 100
GenBank mRNA KA310805.1 - 2 - 3 268 100

View Full Product Details