Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLK10
Entrez Gene ID
102168758
Gene Name
kallikrein related peptidase 10
Gene Aliases
-
Chromosome Location
Chr.18: 58086032 - 58091971 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018062870.1 XP_017918359.1 5 - 6 876 103
XM_018062871.1 XP_017918360.1 5 - 6 806 103
XM_018062872.1 XP_017918361.1 5 - 6 855 103

View Full Product Details