Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HSPB9
Entrez Gene ID
102184756
Gene Name
heat shock protein family B (small) member 9
Gene Aliases
-
Chromosome Location
Chr.19: 41896863 - 41897420 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005709618.3 XP_005709675.2 1 - 1 478 90
GenBank mRNA JX088726.2 - 1 - 1 425 90

View Full Product Details