Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NAIP
Entrez Gene ID
102185222
Gene Name
NLR family apoptosis inhibitory protein
Gene Aliases
-
Chromosome Location
Chr.20: 10150669 - 10193624 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005694594.3 XP_005694651.2 3 - 4 779 86

View Full Product Details