Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
C11H2orf40
Entrez Gene ID
102180322
Gene Name
chromosome 11 C2orf40 homolog
Gene Aliases
-
Chromosome Location
Chr.11: 45405923 - 45415887 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018055322.1 XP_017910811.1 2 - 3 803 114
GenBank mRNA JT701172.1 - 3 - 4 457 114

View Full Product Details