Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PIGH
Entrez Gene ID
102170792
Gene Name
phosphatidylinositol glycan anchor biosynthesis class H
Gene Aliases
-
Chromosome Location
Chr.10: 23685009 - 23692118 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005685998.3 XP_005686055.2 3 - 4 651 89
GenBank mRNA JO438303.2 - 3 - 4 567 89
KA315168.1 - 1 - 2 649 89

View Full Product Details