Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDKN2AIPNL
Entrez Gene ID
102180759
Gene Name
CDKN2A interacting protein N-terminal like
Gene Aliases
-
Chromosome Location
Chr.7: 64533477 - 64539905 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005683309.3 XP_005683366.2 2 - 3 538 100
GenBank mRNA JO612364.2 - 1 - 2 471 100
KA345189.1 - 2 - 3 419 100

View Full Product Details