Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MMRN2
Entrez Gene ID
102172035
Gene Name
multimerin 2
Gene Aliases
-
Chromosome Location
Chr.28: 4150878 - 4166014 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018042186.1 XP_017897675.1 4 - 5 547 102
GenBank mRNA JO415925.2 - 4 - 5 550 102

View Full Product Details