Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ptgr1
Entrez Gene ID
100135572
Gene Name
prostaglandin reductase 1
Gene Aliases
Ltb4dh
Chromosome Location
Chr.Un|NT_176326: 3649185 - 3672490 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172980.1 NP_001166451.1 6 - 7 698 94
GenBank mRNA AB021219.1 - 6 - 7 698 94

View Full Product Details