Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Pol32
- Entrez Gene ID
-
34892
- Gene Name
- Pol32 polymerase delta subunit
- Gene Aliases
-
BG:DS07295.3, BcDNA:GH08865, CG3975, Dmel\CG3975, Dmel_CG3975, pol32, rd
- Chromosome Location
-
Chr.2L: 15254819 - 15256304 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0283467
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF145654.1 |
- |
1 - 1 |
3529 |
64 |
View Full Product Details