Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mkp3
Entrez Gene ID
40081
Gene Name
Mitogen-activated protein kinase phosphatase 3
Gene Aliases
CG14080, CG14081, DMKP-3, DMKP3, Dmel\CG14080, Dmel_CG14080, Dmkp3, MKP3, dMKP3, mkp-3, mkp3
Chromosome Location
Chr.3L: 19066155 - 19086426 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0036844
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275102.1 NP_001262031.1 3 - 4 1437 82
NM_001275103.1 NP_001262032.1 3 - 4 1437 82
NM_140830.2 NP_649087.1 3 - 4 1437 82
NM_168785.1 NP_730385.1 3 - 4 371 82

View Full Product Details