Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lis-1
Entrez Gene ID
36791
Gene Name
Lissencephaly-1
Gene Aliases
CG8440, D-Lis1, DLIS-1, DLIS1, DLis-1, DLis1, Dlis-1, Dlis1, DmLIS1, Dmel\CG8440, Dmel_CG8440, LIS-1, Lis, Lis 1, Lis1, dLis1, l(2)k13209, l(2R)W8, lis-1, lis1, n(2)k11702
Chromosome Location
Chr.2R: 16178826 - 16185103 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0015754
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259432.2 NP_001246361.1 4 - 5 625 90
NM_057812.5 NP_477160.1 4 - 5 700 90
NM_176190.2 NP_788370.1 4 - 5 613 90
NM_206130.2 NP_995852.1 4 - 5 1130 90
GenBank mRNA AF117606.1 - 4 - 5 704 90
AF152419.1 - 4 - 5 689 90
BT021404.1 - 4 - 5 695 90

View Full Product Details