Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lmpt
Entrez Gene ID
39889
Gene Name
Limpet
Gene Aliases
BcDNA:RE37250, CG11914, CG11915, CG11916, CG32171, CG33256, CG42679, CG9959, Dmel\CG42679, Dmel_CG42679, lmpt
Chromosome Location
Chr.3L: 16865295 - 16922487 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261565
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_170635.3 NP_730212.1 2 - 3 456 110
NM_206387.3 NP_996109.1 3 - 4 400 110
GenBank mRNA BT100317.1 - 2 - 3 454 110

View Full Product Details