Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lmpt
Entrez Gene ID
39889
Gene Name
Limpet
Gene Aliases
BcDNA:RE37250, CG11914, CG11915, CG11916, CG32171, CG33256, CG42679, CG9959, Dmel\CG42679, Dmel_CG42679, lmpt
Chromosome Location
Chr.3L: 16865295 - 16922487 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261565
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202193.2 NP_001189122.1 3 - 4 2917 64
NM_001259879.2 NP_001246808.1 6 - 7 1321 64
NM_001275047.1 NP_001261976.2 4 - 5 1106 64
NM_001316485.1 NP_001303414.1 2 - 3 310 64
NM_140673.5 NP_648930.1 6 - 7 1332 64
NM_168694.3 NP_730213.1 2 - 3 515 64
NM_170635.3 NP_730212.1 5 - 6 1276 64
NM_206387.3 NP_996109.1 6 - 7 1220 64
NM_206388.3 NP_996110.1 6 - 7 1444 64
NM_206389.3 NP_996111.1 2 - 3 526 64
GenBank mRNA AJ418713.1 - 6 - 7 1314 64
AY069696.1 - 6 - 7 1314 64
BT009969.1 - 6 - 7 1329 64
BT044112.1 - 2 - 3 514 64
BT100317.1 - 5 - 6 1274 64

View Full Product Details