Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Doc2
Entrez Gene ID
39038
Gene Name
Dorsocross2
Gene Aliases
CG5187, Dm-DOC2, Dmel\CG5187, Dmel_CG5187, Doc, Doc-2, doc2
Chromosome Location
Chr.3L: 9012686 - 9019246 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0035956
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_140025.4 NP_648282.1 2 - 3 605 72
GenBank mRNA AF444796.1 - 2 - 3 599 72
AF444797.1 - 4 - 5 1171 72
AY084173.1 - 2 - 3 599 72

View Full Product Details