The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001259054.2 | NP_001245983.1 | 6 - 7 | 4408 | 52 |
NM_078824.3 | NP_523548.1 | 6 - 7 | 3600 | 52 | |
NM_164965.2 | NP_723669.1 | 6 - 7 | 3866 | 52 | |
GenBank mRNA | BT150428.1 | - | 6 - 7 | 3837 | 52 |