Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- mop
- Entrez Gene ID
-
39610
- Gene Name
- myopic
- Gene Aliases
-
CG9311, Dmel\CG9311, Dmel_CG9311, MOP, Mop, sfv
- Chromosome Location
-
Chr.3L: 14769341 - 14776129 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0036448
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_140465.3 |
NP_648722.2 |
4 - 5 |
4906 |
86 |
View Full Product Details