Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BicC
Entrez Gene ID
34946
Gene Name
Bicaudal C
Gene Aliases
BG:DS00913.2, BICC, Bic-C, CG4824, Dmel\CG4824, Dmel_CG4824, Dromel_CG4824_FBtr0080804_bicc_mORF, bicC, br34, l(2)br34
Chromosome Location
Chr.2L: 16042047 - 16048757 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0000182
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057517.3 NP_476865.1 5 - 6 1312 68
NM_165144.2 NP_723948.1 4 - 5 989 68
NM_165145.1 NP_723949.1 1 - 2 308 68
GenBank mRNA BT023837.1 - 5 - 6 1229 68

Gene Symbol
CG45084
Entrez Gene ID
19836008
Gene Name
CG45084-PA
Gene Aliases
Dmel\CG45084, Dmel_CG45084, Dromel_CG4824_FBtr0080804_bicc_uORF
Chromosome Location
Chr.2L: 16042047 - 16048757 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0266459
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299044.2 NP_001285973.1 4 - 5 989 68
NM_001299045.1 NP_001285974.1 5 - 6 1312 68

View Full Product Details