Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hcf
Entrez Gene ID
43788
Gene Name
Host cell factor
Gene Aliases
CG1710, Dmel\CG1710, Dmel_CG1710, HCF, HCF1, HCF_DROME, Hcf1, anon-WO0172774.48, dHCF, dHcf, dHcf1
Chromosome Location
Chr.4: 359938 - 375554 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0039904
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258491.3 NP_001245420.1 4 - 5 1362 73
NM_079882.4 NP_524621.2 4 - 5 1288 73
NM_166756.3 NP_726566.1 4 - 5 984 73
NM_166757.3 NP_726567.2 4 - 5 1288 73
NM_205873.3 NP_995595.1 4 - 5 984 73
GenBank mRNA AJ320236.1 - 3 - 4 941 73
BT015268.1 - 4 - 5 1249 73

View Full Product Details