Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tadr
Entrez Gene ID
35370
Gene Name
torn and diminished rhabdomeres
Gene Aliases
CG9264, Dmel\CG9264, Dmel_CG9264, torn
Chromosome Location
Chr.2L: 21075405 - 21081053 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0032911
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_136238.3 NP_610082.2 2 - 3 459 59
NM_165354.2 NP_724308.2 2 - 3 459 59

View Full Product Details