Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pkcdelta
Entrez Gene ID
32191
Gene Name
Protein kinase C delta
Gene Aliases
CG10524, CG11245, CG42349, Dmel\CG42349, Dmel_CG42349, PKCd, PKCdelta
Chromosome Location
Chr.X: 12432224 - 12453650 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0259680
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144719.2 NP_001138191.1 6 - 7 1290 60
NM_078616.4 NP_511171.3 6 - 7 1466 60
GenBank mRNA BT021275.1 - 5 - 6 1222 60

View Full Product Details