Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Or43a
- Entrez Gene ID
-
35644
- Gene Name
- Odorant receptor 43a
- Gene Aliases
-
43B.1, 43B1, 43a, AN14, CG1854, DOR43B.1, DOR43a, DOR87, Dmel\CG1854, Dmel_CG1854, Dor87, OR43a, Or, Or43A, Or43B.1, Or87, dor87, or43a
- Chromosome Location
-
Chr.2R: 7232840 - 7234913 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0026389
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078923.3 |
NP_523647.2 |
3 - 4 |
830 |
97 |
View Full Product Details