Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Ance-4
- Entrez Gene ID
-
35909
- Gene Name
- Ance-4-PA
- Gene Aliases
-
CG8196, Dmel\CG8196, Dmel_CG8196, ance-4
- Chromosome Location
-
Chr.2R: 9039728 - 9042142 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0033366
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_136598.3 |
NP_610442.2 |
1 - 2 |
103 |
69 |
View Full Product Details