Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Prp8
- Entrez Gene ID
-
36304
- Gene Name
- pre-mRNA processing factor 8
- Gene Aliases
-
A1Z8U0, CG8877, Dmel\CG8877, Dmel_CG8877, E-2e, prp8
- Chromosome Location
-
Chr.2R: 12154258 - 12162998 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0033688
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_136891.4 |
NP_610735.1 |
12 - 13 |
4091 |
60 |
View Full Product Details